The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077484.1 | NP_001070952.1 | 6 - 7 | 1150 | 150 |
NM_001278387.1 | NP_001265316.1 | 7 - 8 | 1612 | 150 | |
NM_001278388.1 | NP_001265317.1 | 5 - 6 | 1374 | 150 | |
NM_001278389.1 | NP_001265318.1 | 6 - 7 | 639 | 150 | |
NM_001278390.1 | NP_001265319.1 | 6 - 7 | 1352 | 150 | |
NM_030674.3 | NP_109599.3 | 6 - 7 | 1489 | 150 | |
XM_011538784.2 | XP_011537086.1 | 6 - 7 | 874 | 150 | |
XM_011538786.2 | XP_011537088.1 | 6 - 7 | 874 | 150 | |
XM_011538787.2 | XP_011537089.1 | 6 - 7 | 874 | 150 | |
XM_017019990.1 | XP_016875479.1 | 6 - 7 | 1104 | 150 | |
GenBank mRNA | AF247166.1 | - | 6 - 7 | 835 | 150 |
AF271070.1 | - | 6 - 7 | 733 | 150 | |
AK027825.1 | - | 6 - 7 | 639 | 150 | |
AK074797.1 | - | 3 - 4 | 208 | 150 | |
AK074949.1 | - | 4 - 5 | 397 | 150 | |
AK225639.1 | - | 6 - 7 | 727 | 150 | |
AK225778.1 | - | 6 - 7 | 749 | 150 | |
AK308129.1 | - | 4 - 5 | 438 | 150 | |
AK309410.1 | - | 6 - 7 | 1066 | 150 | |
AK313731.1 | - | 4 - 5 | 440 | 150 | |
BC010620.1 | - | 6 - 7 | 696 | 150 | |
BX647547.1 | - | 7 - 8 | 1184 | 150 | |
BX648495.1 | - | 6 - 7 | 1077 | 150 | |
BX648676.1 | - | 5 - 6 | 944 | 150 |