The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002543.3 | NP_002534.1 | 4 - 5 | 674 | 118 |
GenBank mRNA | AB010710.1 | - | 4 - 5 | 621 | 118 |
AB102861.1 | - | 4 - 5 | 560 | 118 | |
AF035776.1 | - | 4 - 5 | 606 | 118 | |
AK292124.1 | - | 4 - 5 | 615 | 118 | |
AK298040.1 | - | 4 - 5 | 484 | 118 | |
AX429248.1 | - | 4 - 5 | 621 | 118 | |
BC022295.1 | - | 4 - 5 | 615 | 118 |