The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021922.2 | NP_068741.1 | 5 - 6 | 1296 | 58 |
XM_005248885.2 | XP_005248942.1 | 5 - 6 | 1297 | 58 | |
XM_005248886.2 | XP_005248943.1 | 4 - 5 | 1249 | 58 | |
XM_005248887.2 | XP_005248944.1 | 5 - 6 | 1318 | 58 | |
XM_005248888.3 | XP_005248945.1 | 5 - 6 | 1318 | 58 | |
XM_011514343.2 | XP_011512645.1 | 5 - 6 | 1011 | 58 | |
GenBank mRNA | AF265210.1 | - | 5 - 6 | 1296 | 58 |
AK292522.1 | - | 5 - 6 | 1307 | 58 | |
AK312617.1 | - | 5 - 6 | 1263 | 58 | |
BC046359.1 | - | 5 - 6 | 1302 | 58 |