The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267895.1 | NP_001254824.1 | 6 - 7 | 707 | 90 |
NM_133455.3 | NP_597712.2 | 6 - 7 | 713 | 90 | |
XM_005261328.2 | XP_005261385.1 | 6 - 7 | 708 | 90 | |
XM_005261329.2 | XP_005261386.1 | 5 - 6 | 618 | 90 | |
GenBank mRNA | AJ416090.1 | - | 6 - 7 | 580 | 90 |
BC046358.1 | - | 6 - 7 | 661 | 90 | |
CR456407.1 | - | 6 - 7 | 596 | 90 |