The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323623.1 | NP_001310552.1 | 15 - 16 | 2319 | 117 |
NM_001323624.1 | NP_001310553.1 | 13 - 14 | 1930 | 117 | |
NM_013285.2 | NP_037417.1 | 14 - 15 | 2115 | 117 | |
GenBank mRNA | AK130697.1 | - | 2 - 3 | 2128 | 117 |
AK312915.1 | - | 14 - 15 | 2069 | 117 | |
BC000107.2 | - | 14 - 15 | 2051 | 117 | |
BC009250.2 | - | 14 - 15 | 2051 | 117 | |
L05425.1 | - | 14 - 15 | 2031 | 117 |