The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145807.2 | NP_665806.1 | 5 - 6 | 1196 | 66 |
XM_011526443.2 | XP_011524745.1 | 5 - 6 | 1234 | 66 | |
XM_017026274.1 | XP_016881763.1 | 4 - 5 | 1422 | 66 | |
GenBank mRNA | BC018654.2 | - | 5 - 6 | 1122 | 66 |
BC018697.2 | - | 5 - 6 | 1122 | 66 | |
BC033207.1 | - | 5 - 6 | 1122 | 66 | |
BE270224.1 | - | 4 - 5 | 545 | 65 |