Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTN5
Entrez Gene ID
126147
Gene Name
netrin 5
Gene Aliases
-
Chromosome Location
Chr.19: 48661407 - 48673119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145807.2 NP_665806.1 6 - 7 1272 66
XM_017026274.1 XP_016881763.1 5 - 6 1498 66
GenBank mRNA BC018654.2 - 6 - 7 1198 66
BC018697.2 - 6 - 7 1198 66
BC033207.1 - 6 - 7 1198 66

View Full Product Details