Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr.9: 17906574 - 18910950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040272.5 NP_001035362.3 25 - 26 4793 70
XM_011518063.2 XP_011516365.1 27 - 28 4931 70
XM_011518064.2 XP_011516366.1 26 - 27 5046 70
XM_011518067.1 XP_011516369.1 17 - 18 3661 70
XM_011518068.2 XP_011516370.1 15 - 16 4264 70
XM_011518070.2 XP_011516372.1 15 - 16 3429 70
XM_017015310.1 XP_016870799.1 26 - 27 11568 70
XM_017015311.1 XP_016870800.1 26 - 27 4880 70
XM_017015312.1 XP_016870801.1 25 - 26 4996 70
XM_017015313.1 XP_016870802.1 26 - 27 4694 70
GenBank mRNA AK123028.1 - 2 - 3 278 70
BC100788.2 - 4 - 5 842 70
BC100790.2 - 4 - 5 841 70
BX640951.1 - 6 - 7 926 70

View Full Product Details