Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
-
Chromosome Location
Chr.11: 74840910 - 74949187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270380.1 NP_001257309.1 11 - 12 1424 143
NM_001270381.1 NP_001257310.1 11 - 12 1417 143
NM_182969.2 NP_892014.1 14 - 15 1685 143
XM_005273765.2 XP_005273822.1 14 - 15 1709 143
XM_005273770.2 XP_005273827.1 12 - 13 1489 143
XM_011544755.1 XP_011543057.1 14 - 15 1664 143
XM_011544756.1 XP_011543058.1 13 - 14 1583 143
XM_011544757.1 XP_011543059.1 13 - 14 1538 143
XM_011544758.1 XP_011543060.1 12 - 13 1538 143
XM_011544759.1 XP_011543061.1 12 - 13 1514 143
XM_011544760.2 XP_011543062.1 14 - 15 1709 143
XM_011544761.1 XP_011543063.1 11 - 12 1466 143
XM_011544762.1 XP_011543064.1 11 - 12 1448 143
XM_011544763.1 XP_011543065.1 14 - 15 1709 143
XM_011544765.1 XP_011543067.1 12 - 13 1423 143
XM_011544766.2 XP_011543068.1 14 - 15 1728 143
XM_011544767.1 XP_011543069.1 13 - 14 1630 143
XM_011544768.1 XP_011543070.1 12 - 13 1477 143
XM_017017222.1 XP_016872711.1 13 - 14 1583 143
XM_017017223.1 XP_016872712.1 12 - 13 1538 143
XM_017017224.1 XP_016872713.1 11 - 12 1448 143
XM_017017226.1 XP_016872715.1 13 - 14 1604 143
XM_017017227.1 XP_016872716.1 13 - 14 1580 143
XM_017017228.1 XP_016872717.1 12 - 13 1489 143
XM_017017229.1 XP_016872718.1 12 - 13 1433 143
XM_017017230.1 XP_016872719.1 11 - 12 1257 143
XM_017017231.1 XP_016872720.1 10 - 11 1343 143
XM_017017233.1 XP_016872722.1 11 - 12 1437 143
XM_017017234.1 XP_016872723.1 10 - 11 1322 143
GenBank mRNA AF439934.1 - 3 - 4 310 143
AK056364.1 - 9 - 10 1008 143
AK074152.1 - 9 - 10 1015 143
BC037294.2 - 11 - 12 1350 143
BC040541.1 - 11 - 12 1350 143
BC064584.1 - 11 - 12 1353 143
CR749605.1 - 10 - 11 1389 143

View Full Product Details