Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
-
Chromosome Location
Chr.11: 74840910 - 74949187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270380.1 NP_001257309.1 13 - 14 1973 65
NM_001270381.1 NP_001257310.1 13 - 14 1966 65
NM_182969.2 NP_892014.1 16 - 17 2234 65
XM_005273765.2 XP_005273822.1 16 - 17 2258 65
XM_005273770.2 XP_005273827.1 14 - 15 2038 65
XM_011544755.1 XP_011543057.1 16 - 17 2213 65
XM_011544756.1 XP_011543058.1 15 - 16 2132 65
XM_011544757.1 XP_011543059.1 15 - 16 2087 65
XM_011544758.1 XP_011543060.1 14 - 15 2087 65
XM_011544759.1 XP_011543061.1 14 - 15 2063 65
XM_011544760.2 XP_011543062.1 16 - 17 2258 65
XM_011544761.1 XP_011543063.1 13 - 14 2015 65
XM_011544762.1 XP_011543064.1 13 - 14 1997 65
XM_011544763.1 XP_011543065.1 16 - 17 2258 65
XM_011544764.1 XP_011543066.1 13 - 14 1922 65
XM_011544765.1 XP_011543067.1 14 - 15 1972 65
XM_011544766.2 XP_011543068.1 16 - 17 2277 65
XM_011544767.1 XP_011543069.1 15 - 16 2179 65
XM_011544768.1 XP_011543070.1 14 - 15 2026 65
XM_017017221.1 XP_016872710.1 14 - 15 2048 65
XM_017017222.1 XP_016872711.1 15 - 16 2132 65
XM_017017223.1 XP_016872712.1 14 - 15 2087 65
XM_017017224.1 XP_016872713.1 13 - 14 1997 65
XM_017017225.1 XP_016872714.1 13 - 14 1922 65
XM_017017226.1 XP_016872715.1 15 - 16 2153 65
XM_017017227.1 XP_016872716.1 15 - 16 2129 65
XM_017017228.1 XP_016872717.1 14 - 15 2038 65
XM_017017229.1 XP_016872718.1 14 - 15 1982 65
XM_017017230.1 XP_016872719.1 13 - 14 1806 65
XM_017017231.1 XP_016872720.1 12 - 13 1892 65
XM_017017232.1 XP_016872721.1 11 - 12 1702 65
XM_017017233.1 XP_016872722.1 13 - 14 1986 65
XM_017017234.1 XP_016872723.1 12 - 13 1871 65
GenBank mRNA AF439934.1 - 5 - 6 859 65
AK056364.1 - 11 - 12 1557 65
AK074152.1 - 11 - 12 1564 65
AK091449.1 - 3 - 4 1680 65
BC037294.2 - 13 - 14 1899 65
BC040541.1 - 13 - 14 1899 65
BC064584.1 - 13 - 14 1902 65
CR749605.1 - 12 - 13 1938 65

View Full Product Details