Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
-
Chromosome Location
Chr.11: 74840910 - 74949187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270380.1 NP_001257309.1 4 - 5 606 69
NM_001270381.1 NP_001257310.1 2 - 3 440 69
NM_182969.2 NP_892014.1 4 - 5 606 69
XM_005273765.2 XP_005273822.1 4 - 5 606 69
XM_005273770.2 XP_005273827.1 2 - 3 386 69
XM_011544755.1 XP_011543057.1 4 - 5 606 69
XM_011544756.1 XP_011543058.1 4 - 5 606 69
XM_011544757.1 XP_011543059.1 4 - 5 606 69
XM_011544758.1 XP_011543060.1 4 - 5 606 69
XM_011544759.1 XP_011543061.1 4 - 5 606 69
XM_011544760.2 XP_011543062.1 4 - 5 606 69
XM_011544762.1 XP_011543064.1 4 - 5 606 69
XM_011544763.1 XP_011543065.1 4 - 5 606 69
XM_011544764.1 XP_011543066.1 4 - 5 606 69
XM_011544765.1 XP_011543067.1 2 - 3 344 69
XM_011544766.2 XP_011543068.1 3 - 4 501 69
XM_011544767.1 XP_011543069.1 3 - 4 501 69
XM_011544768.1 XP_011543070.1 2 - 3 348 69
XM_017017221.1 XP_016872710.1 4 - 5 606 69
XM_017017222.1 XP_016872711.1 4 - 5 606 69
XM_017017223.1 XP_016872712.1 4 - 5 606 69
XM_017017224.1 XP_016872713.1 4 - 5 606 69
XM_017017225.1 XP_016872714.1 4 - 5 606 69
XM_017017226.1 XP_016872715.1 3 - 4 501 69
XM_017017227.1 XP_016872716.1 3 - 4 501 69
XM_017017228.1 XP_016872717.1 2 - 3 386 69
XM_017017229.1 XP_016872718.1 3 - 4 501 69
XM_017017230.1 XP_016872719.1 3 - 4 501 69
XM_017017231.1 XP_016872720.1 3 - 4 501 69
XM_017017232.1 XP_016872721.1 2 - 3 386 69
XM_017017233.1 XP_016872722.1 3 - 4 501 69
XM_017017234.1 XP_016872723.1 2 - 3 386 69
GenBank mRNA AK056364.1 - 3 - 4 388 69
AK074152.1 - 2 - 3 222 69
BC037294.2 - 2 - 3 373 69
BC040541.1 - 2 - 3 373 69
BC064584.1 - 4 - 5 535 69
CR749605.1 - 2 - 3 453 69
DB040217.1 - 4 - 5 522 69

View Full Product Details