The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005204.3 | NP_005195.2 | 1 - 2 | 363 | 111 |
XM_017015713.1 | XP_016871202.1 | 1 - 2 | 337 | 111 | |
GenBank mRNA | AL547407.3 | - | 1 - 2 | 454 | 111 |
D14497.1 | - | 1 - 2 | 119 | 110 | |
DA308072.1 | - | 1 - 2 | 337 | 111 |