The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181805.2 | NP_861521.1 | 3 - 4 | 451 | 97 |
XM_017027614.1 | XP_016883103.1 | 2 - 3 | 330 | 97 | |
GenBank mRNA | BG717090.1 | - | 3 - 4 | 365 | 97 |