The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184699.1 | NP_001171628.1 | 5 - 6 | 755 | 100 |
NM_019609.4 | NP_062555.1 | 5 - 6 | 755 | 100 | |
GenBank mRNA | AK027661.1 | - | 5 - 6 | 691 | 100 |
AK075527.1 | - | 5 - 6 | 730 | 100 | |
AK303142.1 | - | 2 - 3 | 234 | 100 | |
AK315027.1 | - | 5 - 6 | 742 | 100 | |
AY358956.1 | - | 5 - 6 | 711 | 100 | |
BC063430.1 | - | 5 - 6 | 732 | 100 |