Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAB2IP
Entrez Gene ID
153090
Gene Name
DAB2 interacting protein
Gene Aliases
AF9Q34, AIP-1, AIP1, DIP1/2
Chromosome Location
Chr.9: 121566883 - 121785530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032552.3 NP_115941.2 1 - 2 130 112
XM_005251721.1 XP_005251778.1 1 - 2 349 112
GenBank mRNA AK096391.1 - 1 - 2 350 112
BM545676.1 - 1 - 2 130 112
BM546615.1 - 1 - 2 112 112

View Full Product Details