Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBNL1
Entrez Gene ID
4154
Gene Name
muscleblind like splicing regulator 1
Gene Aliases
EXP, MBNL
Chromosome Location
Chr.3: 152243656 - 152465780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314057.1 NP_001300986.1 6 - 7 1433 93
NM_207293.1 NP_997176.1 4 - 5 1593 93
XM_005247457.4 XP_005247514.1 6 - 7 1786 93
XM_005247458.4 XP_005247515.1 6 - 7 3040 93
XM_005247460.4 XP_005247517.1 6 - 7 1749 93
XM_005247461.4 XP_005247518.1 6 - 7 1786 93
XM_005247464.4 XP_005247521.1 5 - 6 1844 93
XM_005247465.4 XP_005247522.1 6 - 7 1786 93
XM_005247466.4 XP_005247523.1 5 - 6 1821 93
XM_005247467.4 XP_005247524.1 6 - 7 1786 93
XM_005247474.3 XP_005247531.1 4 - 5 704 93
XM_005247475.3 XP_005247532.1 5 - 6 1027 93
XM_005247477.4 XP_005247534.1 4 - 5 793 93
XM_006713639.1 XP_006713702.1 6 - 7 1768 93
XM_011512843.2 XP_011511145.1 5 - 6 1582 93
XM_011512848.2 XP_011511150.1 6 - 7 1390 93
XM_017006423.1 XP_016861912.1 5 - 6 2037 93
XM_017006424.1 XP_016861913.1 5 - 6 1864 93
XM_017006425.1 XP_016861914.1 5 - 6 1866 93
XM_017006426.1 XP_016861915.1 5 - 6 1864 93
XM_017006432.1 XP_016861921.1 5 - 6 1821 93
XM_017006433.1 XP_016861922.1 5 - 6 1821 93
XM_017006434.1 XP_016861923.1 5 - 6 1827 93
XM_017006435.1 XP_016861924.1 5 - 6 834 93
XM_017006438.1 XP_016861927.1 4 - 5 1489 93
GenBank mRNA AK304089.1 - 3 - 4 513 93
BC050535.1 - 6 - 7 954 93
Y13829.1 - 4 - 5 1315 93

View Full Product Details