Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258008.1 NP_001244937.1 7 - 8 1010 103
XM_005250479.2 XP_005250536.1 7 - 8 1441 103
XM_005250480.2 XP_005250537.1 7 - 8 1441 103
XM_006716035.3 XP_006716098.1 7 - 8 1441 103
XM_006716036.2 XP_006716099.1 7 - 8 945 103
XM_006716037.3 XP_006716100.1 7 - 8 1441 103
XM_006716040.3 XP_006716103.1 7 - 8 1441 103
XM_017012379.1 XP_016867868.1 7 - 8 1441 103
XM_017012380.1 XP_016867869.1 7 - 8 945 103
GenBank mRNA AK000919.1 - 2 - 3 394 103
AK093038.1 - 7 - 8 945 103
AK300745.1 - 7 - 8 866 103
AK302108.1 - 6 - 7 744 103
BC002725.1 - 4 - 5 708 103
KU178677.1 - 2 - 3 270 103
KU178678.1 - 2 - 3 270 103
KU178679.1 - 2 - 3 270 103
KU178680.1 - 2 - 3 270 103
KU178681.1 - 2 - 3 270 103
KU178682.1 - 2 - 3 270 103

View Full Product Details