Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPA12A
Entrez Gene ID
259217
Gene Name
heat shock protein family A (Hsp70) member 12A
Gene Aliases
-
Chromosome Location
Chr.10: 116671192 - 116851241 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025015.2 NP_079291.2 8 - 9 1028 94
XM_005269672.3 XP_005269729.1 9 - 10 1178 94
XM_005269673.4 XP_005269730.1 9 - 10 2122 94
XM_011539579.2 XP_011537881.1 10 - 11 1239 94
XM_011539580.2 XP_011537882.1 9 - 10 1184 94
XM_017016032.1 XP_016871521.1 6 - 7 750 94
GenBank mRNA AB007877.1 - 8 - 9 1028 94
AK294672.1 - 7 - 8 842 94
AK294884.1 - 3 - 4 347 94

View Full Product Details