The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000072.3 | NP_000063.2 | 2 - 3 | 324 | 104 |
NM_001001547.2 | NP_001001547.1 | 2 - 3 | 285 | 104 | |
NM_001001548.2 | NP_001001548.1 | 2 - 3 | 324 | 104 | |
NM_001289909.1 | NP_001276838.1 | 1 - 2 | 89 | 104 | |
NR_110501.1 | 1 - 2 | 89 | 104 | ||
XM_005250713.1 | XP_005250770.1 | 2 - 3 | 266 | 104 | |
XM_005250714.1 | XP_005250771.1 | 2 - 3 | 233 | 104 | |
XM_005250715.4 | XP_005250772.1 | 2 - 3 | 391 | 104 | |
GenBank mRNA | AK096858.1 | - | 5 - 6 | 594 | 104 |
AK129899.1 | - | 2 - 3 | 266 | 104 | |
AK312920.1 | - | 2 - 3 | 248 | 104 | |
BC008406.1 | - | 2 - 3 | 199 | 104 | |
BP258762.1 | - | 2 - 3 | 285 | 104 | |
BP259746.1 | - | 2 - 3 | 284 | 104 | |
DA857400.1 | - | 2 - 3 | 104 | ||
DC338512.1 | - | 2 - 3 | 225 | 104 | |
M24795.1 | - | 2 - 3 | 120 | 104 | |
M98398.1 | - | 2 - 3 | 164 | 104 | |
M98399.1 | - | 2 - 3 | 164 | 104 | |
S67532.1 | - | 1 - 2 | 46 | 104 |