The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001151.3 | NP_001142.2 | 2 - 3 | 715 | 86 |
GenBank mRNA | AK291902.1 | - | 2 - 3 | 709 | 86 |
BC008664.1 | - | 2 - 3 | 703 | 86 | |
BC061589.1 | - | 2 - 3 | 721 | 86 | |
BC063643.1 | - | 2 - 3 | 704 | 86 | |
J02966.1 | - | 2 - 3 | 705 | 86 | |
KU177900.1 | - | 2 - 3 | 605 | 86 | |
KU177901.1 | - | 2 - 3 | 239 | 86 |