The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184690.1 | NP_001171619.1 | 2 - 3 | 381 | 63 |
XM_005262888.4 | XP_005262945.1 | 2 - 3 | 368 | 63 | |
XM_011531820.2 | XP_011530122.1 | 2 - 3 | 371 | 63 | |
XM_011531823.2 | XP_011530125.1 | 2 - 3 | 371 | 63 | |
XM_011531824.2 | XP_011530126.1 | 2 - 3 | 371 | 63 | |
XM_017007984.1 | XP_016863473.1 | 2 - 3 | 371 | 63 | |
GenBank mRNA | AK295772.1 | - | 2 - 3 | 381 | 63 |