Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
Chromosome Location
Chr.1: 32651148 - 32686211 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135255.1 NP_001128727.1 11 - 12 1368 112
NM_001135256.1 NP_001128728.1 11 - 12 1271 112
NM_005610.2 NP_005601.1 11 - 12 1371 112
GenBank mRNA AK222779.1 - 11 - 12 1274 112
AK299251.1 - 11 - 12 1271 112
AK299410.1 - 9 - 10 982 112
AK312571.1 - 11 - 12 1309 112
BC003092.1 - 11 - 12 1298 112
BC015123.1 - 9 - 10 1032 112
BC053904.1 - 11 - 12 1284 112
BC075836.1 - 11 - 12 1239 112
BT007309.1 - 11 - 12 1212 112
X74262.1 - 11 - 12 1296 112

View Full Product Details