The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015191.2 | NP_056006.1 | 7 - 8 | 1116 | 73 |
XM_017017417.1 | XP_016872906.1 | 7 - 8 | 1075 | 73 | |
XM_017017418.1 | XP_016872907.1 | 5 - 6 | 645 | 73 | |
GenBank mRNA | AB018324.3 | - | 7 - 8 | 1017 | 73 |
AB084424.1 | - | 7 - 8 | 937 | 73 | |
AB096248.1 | - | 7 - 8 | 1022 | 73 | |
AK291233.1 | - | 7 - 8 | 1051 | 73 | |
AL136764.2 | - | 7 - 8 | 1039 | 73 | |
BC078150.1 | - | 2 - 3 | 299 | 73 | |
BC113459.1 | - | 7 - 8 | 973 | 73 | |
BC117183.1 | - | 7 - 8 | 973 | 73 |