Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2E2
Entrez Gene ID
2961
Gene Name
general transcription factor IIE subunit 2
Gene Aliases
FE, TF2E2, TFIIE-B, TTD6
Chromosome Location
Chr.8: 30578514 - 30658241 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017013364.1 XP_016868853.1 1 - 2 36 64

View Full Product Details