The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002095.4 | NP_002086.1 | 4 - 5 | 612 | 167 |
XM_011544509.2 | XP_011542811.1 | 4 - 5 | 632 | 167 | |
XM_011544510.1 | XP_011542812.1 | 4 - 5 | 632 | 167 | |
XM_017013363.1 | XP_016868852.1 | 4 - 5 | 472 | 167 | |
XM_017013364.1 | XP_016868853.1 | 4 - 5 | 402 | 167 | |
GenBank mRNA | BC030572.2 | - | 4 - 5 | 606 | 167 |
S67861.1 | - | 4 - 5 | 549 | 167 | |
X63469.1 | - | 4 - 5 | 601 | 167 |