Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPC6
Entrez Gene ID
10082
Gene Name
glypican 6
Gene Aliases
OMIMD1
Chromosome Location
Chr.13: 93226825 - 94408020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005708.3 NP_005699.1 2 - 3 948 108
XM_011521044.2 XP_011519346.1 2 - 3 273 108
XM_017020298.1 XP_016875787.1 2 - 3 2556 108
XM_017020299.1 XP_016875788.1 2 - 3 1323 108
XM_017020300.1 XP_016875789.1 2 - 3 180 108
GenBank mRNA AF105267.1 - 2 - 3 901 108
AF111178.1 - 2 - 3 950 108
AK074741.1 - 2 - 3 402 108
AK130013.1 - 2 - 3 285 108
AK290144.1 - 2 - 3 469 108
AK304338.1 - 2 - 3 767 108
AY358462.1 - 2 - 3 881 108
BC106947.2 - 2 - 3 796 108

View Full Product Details