Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PCDHB19P
- Entrez Gene ID
-
84054
- Gene Name
- protocadherin beta 19 pseudogene
- Gene Aliases
-
PCDH-PSI5, PCDHB19
- Chromosome Location
-
Chr.5: 141240121 - 141244752 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001282.2 |
|
1 - 1 |
3638 |
130 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF329369.3 |
- |
2 |
3618 |
|
Chr. 5 - 141240121 - 141244752 [+] on Build GRCh38 |
130 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protocadherin beta 18 pseudogene |
PCDHB18P |
PCDH-psi2, PCDHB18 |
-2.6kb
|
Sense |
Chr. - 141234333 - 141237529 on Build GRCh38 |
Downstream |
protocadherin beta 15 |
PCDHB15 |
PCDH-BETA15 |
597.0
|
Sense |
Chr. - 141245349 - 141248234 on Build GRCh38 |
View Full Product Details