The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021072.3 | NP_066550.2 | 4 - 5 | 1258 | 72 |
GenBank mRNA | AF064876.1 | - | 4 - 5 | 870 | 72 |
AF488549.1 | - | 4 - 5 | 1258 | 72 |