Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNAL2
Entrez Gene ID
83473
Gene Name
katanin catalytic subunit A1 like 2
Gene Aliases
-
Chromosome Location
Chr.18: 46917602 - 47104795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031303.2 NP_112593.2 13 - 14 1342 74
XM_005258357.4 XP_005258414.1 16 - 17 1887 74
XM_005258358.4 XP_005258415.1 16 - 17 1884 74
XM_005258361.3 XP_005258418.1 12 - 13 1144 74
XM_006722554.3 XP_006722617.1 16 - 17 2198 74
XM_017026030.1 XP_016881519.1 15 - 16 1838 74
XM_017026031.1 XP_016881520.1 13 - 14 1323 74
GenBank mRNA BC034999.2 - 13 - 14 1342 74

View Full Product Details