Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNAL2
Entrez Gene ID
83473
Gene Name
katanin catalytic subunit A1 like 2
Gene Aliases
-
Chromosome Location
Chr.18: 46917602 - 47104795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031303.2 NP_112593.2 8 - 9 809 91
XM_005258357.4 XP_005258414.1 11 - 12 1354 91
XM_005258358.4 XP_005258415.1 11 - 12 1351 91
XM_005258361.3 XP_005258418.1 7 - 8 611 91
XM_006722554.3 XP_006722617.1 11 - 12 1665 91
XM_011526219.2 XP_011524521.1 11 - 12 1354 91
XM_011526220.1 XP_011524522.1 11 - 12 1354 91
XM_011526221.2 XP_011524523.1 11 - 12 1354 91
XM_011526223.2 XP_011524525.1 11 - 12 1354 91
XM_017026028.1 XP_016881517.1 11 - 12 1354 91
XM_017026029.1 XP_016881518.1 11 - 12 1665 91
XM_017026030.1 XP_016881519.1 10 - 11 1305 91
XM_017026031.1 XP_016881520.1 8 - 9 790 91
XM_017026032.1 XP_016881521.1 11 - 12 1395 91
GenBank mRNA BC034999.2 - 8 - 9 809 91
CN303333.1 - 5 - 6 512 91

View Full Product Details