Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TDH
- Entrez Gene ID
-
157739
- Gene Name
- L-threonine dehydrogenase (pseudogene)
- Gene Aliases
-
SDR14E1P
- Chromosome Location
-
Chr.8: 11339637 - 11368452 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001578.1 |
|
5 - 6 |
540 |
58 |
GenBank mRNA |
AJ301562.1 |
- |
5 - 6 |
540 |
58 |
AY101186.1 |
- |
4 - 5 |
442 |
58 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DN831681.1 |
- |
2 - 3 |
|
|
Chr. 8 - 11358148 - 11364858 [-] on Build GRCh38 |
58 |
GenBankmRNA |
HY113237.1 |
- |
1 - 2 |
|
|
Chr. 8 - 11361473 - 11365598 [+] on Build GRCh38 |
58 |
GenBankmRNA |
EB710171.1 |
- |
|
|
|
Chr. - - [-] on Build |
58 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 35 member G5 |
SLC35G5 |
AMAC, AMAC1L2 |
-29.2kb
|
Anti-sense |
Chr. - 11330986 - 11332186 on Build GRCh38 |
Downstream |
FAM167A antisense RNA 1 |
FAM167A-AS1 |
C8orf12 |
6.6kb
|
Anti-sense |
Chr. - 11368402 - 11438657 on Build GRCh38 |
View Full Product Details