Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPPA3P2
Entrez Gene ID
400206
Gene Name
developmental pluripotency associated 3 pseudogene 2
Gene Aliases
STELLAR
Chromosome Location
Chr.14: 36371318 - 36372369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC062480.1 - 1 - 1 190 75

View Full Product Details