The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278629.1 | NP_001265558.1 | 9 - 10 | 1268 | 157 |
NM_001278630.1 | NP_001265559.1 | 9 - 10 | 1125 | 157 | |
NM_152786.2 | NP_689999.1 | 9 - 10 | 1341 | 157 | |
XM_005251885.3 | XP_005251942.1 | 7 - 8 | 647 | 157 | |
XM_011518477.1 | XP_011516779.1 | 9 - 10 | 1268 | 157 | |
XM_011518478.1 | XP_011516780.1 | 9 - 10 | 1268 | 157 | |
XM_011518479.2 | XP_011516781.1 | 7 - 8 | 923 | 157 | |
GenBank mRNA | AK301872.1 | - | 9 - 10 | 1125 | 160 |
AK313867.1 | - | 9 - 10 | 1257 | 157 | |
BC026884.1 | - | 9 - 10 | 1307 | 157 | |
BC033037.2 | - | 9 - 10 | 1226 | 157 |