Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FARSB
Entrez Gene ID
10056
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Gene Aliases
FARSLB, FRSB, HSPC173, PheHB, PheRS
Chromosome Location
Chr.2: 222571443 - 222656355 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_130154.1 3 - 4 594 127
XM_006712169.2 XP_006712232.1 3 - 4 300 127

View Full Product Details