The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258008.1 | NP_001244937.1 | 15 - 16 | 1793 | 150 |
XM_005250479.2 | XP_005250536.1 | 15 - 16 | 2227 | 150 | |
XM_005250480.2 | XP_005250537.1 | 14 - 15 | 2146 | 150 | |
XM_017012379.1 | XP_016867868.1 | 14 - 15 | 2143 | 150 | |
XM_017012380.1 | XP_016867869.1 | 14 - 15 | 1647 | 150 | |
GenBank mRNA | AK000919.1 | - | 10 - 11 | 1177 | 150 |
AK300745.1 | - | 15 - 16 | 1649 | 150 | |
KU178680.1 | - | 10 - 11 | 1056 | 150 | |
KU178681.1 | - | 10 - 11 | 1053 | 150 | |
KU178682.1 | - | 9 - 10 | 972 | 150 |