Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACTB
Entrez Gene ID
114294
Gene Name
lactamase beta
Gene Aliases
G24, MRPL56
Chromosome Location
Chr.15: 63121800 - 63142065 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032857.4 NP_116246.2 5 - 6 1183 80
GenBank mRNA BC067288.1 - 5 - 6 1163 80
BG250900.1 - 1 - 2 50 80

View Full Product Details