The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145046.4 | NP_659483.2 | 7 - 8 | 980 | 147 |
GenBank mRNA | AB576176.1 | - | 7 - 8 | 917 | 147 |
AK058084.1 | - | 7 - 8 | 972 | 147 | |
BC014595.2 | - | 7 - 8 | 980 | 147 |