The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015693.3 | NP_056508.2 | 4 - 5 | 1077 | 123 |
XM_011531845.2 | XP_011530147.1 | 4 - 5 | 1104 | 123 | |
XM_011531846.1 | XP_011530148.1 | 4 - 5 | 1049 | 123 | |
XM_011531849.2 | XP_011530151.1 | 4 - 5 | 1104 | 123 | |
XM_011531850.2 | XP_011530152.1 | 4 - 5 | 1104 | 123 | |
XM_011531851.2 | XP_011530153.1 | 4 - 5 | 1104 | 123 | |
XM_017008026.1 | XP_016863515.1 | 4 - 5 | 1104 | 123 | |
GenBank mRNA | AB033110.1 | - | 4 - 5 | 1044 | 123 |
AK291481.1 | - | 4 - 5 | 1059 | 123 | |
BC051698.1 | - | 4 - 5 | 1052 | 123 | |
BC130611.1 | - | 4 - 5 | 1043 | 123 |