The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002591.3 | NP_002582.3 | 4 - 5 | 769 | 104 |
XM_011528839.1 | XP_011527141.1 | 2 - 3 | 411 | 104 | |
GenBank mRNA | AK290802.1 | - | 4 - 5 | 747 | 104 |
AK303806.1 | - | 2 - 3 | 321 | 104 | |
BC023978.1 | - | 4 - 5 | 744 | 104 | |
BJ993061.1 | - | 2 - 3 | 230 | 104 | |
L05144.1 | - | 4 - 5 | 727 | 104 |