Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC2A13
Entrez Gene ID
114134
Gene Name
solute carrier family 2 member 13
Gene Aliases
HMIT
Chromosome Location
Chr.12: 39755021 - 40106085 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052885.3 NP_443117.3 2 - 3 763 74
XM_011537847.2 XP_011536149.1 2 - 3 989 74
XM_011537849.2 XP_011536151.1 2 - 3 989 74
XM_011537850.2 XP_011536152.1 2 - 3 989 74
XM_017018764.1 XP_016874253.1 2 - 3 236 74
XM_017018765.1 XP_016874254.1 2 - 3 874 74
GenBank mRNA BC047507.1 - 2 - 3 993 74
BC117117.1 - 2 - 3 707 74
BC117119.1 - 2 - 3 707 74

View Full Product Details