Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A13
Entrez Gene ID
10165
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
Chromosome Location
Chr.7: 96120220 - 96322147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160210.1 NP_001153682.1 11 - 12 1378 104
NM_014251.2 NP_055066.1 11 - 12 1375 104
NR_027662.1 10 - 11 1259 104
XM_006715831.3 XP_006715894.1 11 - 12 1311 104
XM_011515727.2 XP_011514029.1 12 - 13 1499 104
XM_017011663.1 XP_016867152.1 10 - 11 1226 104
XM_017011664.1 XP_016867153.1 10 - 11 1261 104
XM_017011665.1 XP_016867154.1 9 - 10 1138 104
GenBank mRNA AF118838.1 - 11 - 12 1321 104
AJ496569.1 - 11 - 12 1274 104
AK000766.1 - 11 - 12 1294 104
AK025227.1 - 8 - 9 890 104
AK025779.1 - 6 - 7 731 104
AK222864.1 - 11 - 12 1294 104
AK294629.1 - 10 - 11 1124 104
BC006566.2 - 11 - 12 1257 104
Y17571.1 - 11 - 12 1224 104

View Full Product Details