The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160210.1 | NP_001153682.1 | 14 - 15 | 1645 | 86 |
NM_014251.2 | NP_055066.1 | 14 - 15 | 1642 | 86 | |
NR_027662.1 | 13 - 14 | 1526 | 86 | ||
XM_006715831.3 | XP_006715894.1 | 14 - 15 | 1578 | 86 | |
XM_017011663.1 | XP_016867152.1 | 13 - 14 | 1493 | 86 | |
XM_017011664.1 | XP_016867153.1 | 13 - 14 | 1528 | 86 | |
XM_017011665.1 | XP_016867154.1 | 12 - 13 | 1405 | 86 | |
GenBank mRNA | AF118838.1 | - | 14 - 15 | 1588 | 86 |
AJ496569.1 | - | 14 - 15 | 1541 | 86 | |
AK000766.1 | - | 14 - 15 | 1561 | 86 | |
AK025227.1 | - | 11 - 12 | 1157 | 86 | |
AK025779.1 | - | 9 - 10 | 998 | 86 | |
AK222864.1 | - | 14 - 15 | 1561 | 86 | |
AK294629.1 | - | 13 - 14 | 1391 | 86 | |
BC006566.2 | - | 14 - 15 | 1524 | 86 | |
Y17571.1 | - | 14 - 15 | 1491 | 86 |