The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004246.2 | NP_004237.1 | 12 - 13 | 1845 | 67 |
XM_005256861.3 | XP_005256918.1 | 10 - 11 | 1021 | 67 | |
XM_011524077.2 | XP_011522379.1 | 13 - 14 | 1959 | 67 | |
XM_017025339.1 | XP_016880828.1 | 12 - 13 | 1902 | 67 | |
XM_017025340.1 | XP_016880829.1 | 10 - 11 | 1678 | 67 | |
XM_017025341.1 | XP_016880830.1 | 9 - 10 | 1571 | 67 | |
GenBank mRNA | AK295765.1 | - | 12 - 13 | 1224 | 67 |
AK314269.1 | - | 12 - 13 | 1845 | 67 | |
BC041176.1 | - | 9 - 10 | 901 | 67 | |
BC043390.1 | - | 10 - 11 | 1040 | 67 | |
BC096261.1 | - | 12 - 13 | 1332 | 67 | |
BC096262.1 | - | 12 - 13 | 1332 | 67 | |
BC096263.1 | - | 12 - 13 | 1328 | 67 | |
BC096264.1 | - | 12 - 13 | 1332 | 67 |