The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004246.2 | NP_004237.1 | 9 - 10 | 1569 | 140 |
XM_005256861.3 | XP_005256918.1 | 7 - 8 | 745 | 140 | |
XM_011524077.2 | XP_011522379.1 | 10 - 11 | 1683 | 140 | |
XM_017025339.1 | XP_016880828.1 | 9 - 10 | 1626 | 140 | |
XM_017025340.1 | XP_016880829.1 | 7 - 8 | 1402 | 140 | |
XM_017025341.1 | XP_016880830.1 | 6 - 7 | 1295 | 140 | |
GenBank mRNA | AF105367.1 | - | 9 - 10 | 1056 | 140 |
AK295765.1 | - | 9 - 10 | 948 | 140 | |
AK314269.1 | - | 9 - 10 | 1569 | 140 | |
BC041176.1 | - | 6 - 7 | 625 | 140 | |
BC043390.1 | - | 7 - 8 | 764 | 140 | |
BC096261.1 | - | 9 - 10 | 1056 | 140 | |
BC096262.1 | - | 9 - 10 | 1056 | 140 | |
BC096263.1 | - | 9 - 10 | 1052 | 140 | |
BC096264.1 | - | 9 - 10 | 1056 | 140 |