The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011534069.1 | XP_011532371.1 | 3 - 3 | 2293 | 133 |
GenBank mRNA | X95583.1 | - | 1 - 1 | 1443 | 144 |