Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
Chromosome Location
Chr.16: 58113588 - 58129425 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013242.2 NP_037374.1 4 - 5 741 109
GenBank mRNA AF087845.1 - 4 - 5 721 109
AF093680.1 - 4 - 5 720 109
AK027099.1 - 4 - 5 662 109
BC005152.2 - 4 - 5 710 109
BU730963.1 - 2 - 3 109

View Full Product Details