The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010906.1 | NP_001010906.1 | 4 - 5 | 407 | 71 |
XM_011544523.2 | XP_011542825.1 | 4 - 5 | 676 | 71 | |
XM_011544524.2 | XP_011542826.1 | 4 - 5 | 2182 | 71 | |
XM_011544526.2 | XP_011542828.1 | 4 - 5 | 676 | 71 | |
XM_017013403.1 | XP_016868892.1 | 4 - 5 | 676 | 71 | |
GenBank mRNA | AB075870.1 | - | 4 - 5 | 408 | 71 |