The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005588.2 | NP_005579.2 | 2 - 3 | 142 | 102 |
XM_011514629.1 | XP_011512931.1 | 2 - 3 | 142 | 102 | |
GenBank mRNA | AF478685.1 | - | 2 - 3 | 110 | 102 |
AK225212.1 | - | 2 - 3 | 132 | 102 | |
AK290282.1 | - | 2 - 3 | 110 | 102 | |
AK315246.1 | - | 2 - 3 | 114 | 102 | |
AK315840.1 | - | 2 - 3 | 112 | 102 | |
BC131714.1 | - | 2 - 3 | 113 | 102 | |
DA432559.1 | - | 2 - 3 | 142 | 102 |