The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005588.2 | NP_005579.2 | 5 - 6 | 304 | 68 |
XM_011514628.1 | XP_011512930.1 | 4 - 5 | 361 | 68 | |
XM_011514629.1 | XP_011512931.1 | 5 - 6 | 304 | 68 | |
GenBank mRNA | AF478685.1 | - | 5 - 6 | 272 | 68 |
AK225212.1 | - | 5 - 6 | 294 | 68 | |
AK290282.1 | - | 5 - 6 | 272 | 68 | |
AK315246.1 | - | 5 - 6 | 276 | 68 | |
AK315840.1 | - | 5 - 6 | 274 | 68 | |
BC131714.1 | - | 5 - 6 | 275 | 68 | |
BC143650.1 | - | 3 - 4 | 443 | 68 | |
BC143651.1 | - | 4 - 5 | 354 | 68 | |
DA432559.1 | - | 5 - 6 | 304 | 68 | |
M82962.1 | - | 5 - 6 | 271 | 68 |