Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEP1A
Entrez Gene ID
4224
Gene Name
meprin A subunit alpha
Gene Aliases
PPHA
Chromosome Location
Chr.6: 46793357 - 46845987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005588.2 NP_005579.2 6 - 7 424 119
XM_011514628.1 XP_011512930.1 5 - 6 481 119
XM_011514629.1 XP_011512931.1 6 - 7 424 119
GenBank mRNA AF478685.1 - 6 - 7 392 119
AK225212.1 - 6 - 7 414 119
AK290282.1 - 6 - 7 392 119
AK315246.1 - 6 - 7 396 119
AK315840.1 - 6 - 7 394 119
BC131714.1 - 6 - 7 395 119
BC143650.1 - 4 - 5 563 119
BC143651.1 - 5 - 6 474 119
DA432559.1 - 6 - 7 424 119
M82962.1 - 6 - 7 391 119

View Full Product Details